Assay Details
| Gene Symbol: XRN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333652.1 | NP_001321327.1 | 18 - 19 | 2341 | 137 | |
| NM_104327.4 | NP_175851.1 | 18 - 19 | 2136 | 137 | ||
| GenBank mRNA | AF286718.1 | - | 18 - 19 | 2055 | 137 | |
| AY064012.1 | - | 18 - 19 | 2052 | 137 | ||
| AY091411.1 | - | 18 - 19 | 1945 | 137 | ||
| BT026022.1 | - | 18 - 19 | 2031 | 137 | ||
More Information
Set Membership: |
|

XRN4
Genomic Map