Assay Details
| Gene Symbol: XRN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333652.1 | NP_001321327.1 | 20 - 21 | 2526 | 90 | |
| NM_104327.4 | NP_175851.1 | 20 - 21 | 2321 | 90 | ||
| GenBank mRNA | AF286718.1 | - | 20 - 21 | 2240 | 90 | |
| AY064012.1 | - | 20 - 21 | 2237 | 90 | ||
| AY091411.1 | - | 20 - 21 | 2130 | 90 | ||
| BT026022.1 | - | 20 - 21 | 2216 | 90 | ||
More Information
Set Membership: |
|

XRN4
Genomic Map