Assay Details
| Gene Symbol: AXR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104373.2 | NP_564672.2 | 1 - 2 | 1248 | 109 | |
| GenBank mRNA | AY074334.1 | - | 1 - 2 | 1248 | 109 | |
| AY122984.1 | - | 1 - 2 | 1202 | 109 | ||
More Information
Set Membership: |
|

AXR4
Genomic Map