Assay Details
Gene Symbol: LOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104376.3 | NP_175900.1 | 2 - 3 | 632 | 75 | |
GenBank mRNA | AY093104.1 | - | 2 - 3 | 576 | 75 | |
BT010358.1 | - | 2 - 3 | 488 | 75 | ||
L04637.1 | - | 2 - 3 | 568 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
