Assay Details
| Gene Symbol: AT1G55090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104383.4 | NP_175906.1 | 13 - 14 | 2222 | 77 | |
| GenBank mRNA | AK228292.1 | - | 13 - 14 | 2137 | 77 | |
| BT010741.1 | - | 12 - 13 | 1979 | 77 | ||
More Information
Set Membership: |
|

AT1G55090
Genomic Map