Assay Details
| Gene Symbol: AT1G55090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104383.4 | NP_175906.1 | 6 - 7 | 1207 | 108 | |
| GenBank mRNA | AK228292.1 | - | 6 - 7 | 1122 | 108 | |
| BT010741.1 | - | 5 - 6 | 964 | 108 | ||
More Information
Set Membership: |
|

AT1G55090
Genomic Map