Assay Details
| Gene Symbol: AT1G55090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104383.4 | NP_175906.1 | 7 - 8 | 1378 | 64 | |
| GenBank mRNA | AK228292.1 | - | 7 - 8 | 1293 | 64 | |
| BT010741.1 | - | 6 - 7 | 1135 | 64 | ||
More Information
Set Membership: |
|

AT1G55090
Genomic Map