Assay Details
Gene Symbol: FRUCT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124022.2 | NP_001117494.1 | 2 - 3 | 1149 | 99 | |
NM_001198301.1 | NP_001185230.1 | 2 - 3 | 1028 | 99 | ||
NM_001333690.1 | NP_001320753.1 | 2 - 3 | 1325 | 99 | ||
NM_104385.4 | NP_564676.1 | 3 - 4 | 1169 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
