Assay Details
| Gene Symbol: RH20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333691.1 | NP_001322402.1 | 3 - 4 | 1021 | 68 | |
| NM_104388.4 | NP_175911.1 | 4 - 5 | 825 | 68 | ||
| GenBank mRNA | AY062745.1 | - | 4 - 5 | 821 | 68 | |
| AY128786.1 | - | 4 - 5 | 671 | 68 | ||
More Information
Set Membership: |
|

RH20
Genomic Map