Assay Details
| Gene Symbol: CPN60B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333710.1 | NP_001320639.1 | 2 - 3 | 654 | 82 | |
| NM_001333711.1 | NP_001320638.1 | 2 - 3 | 637 | 82 | ||
| NM_001333712.1 | NP_001320637.1 | 2 - 3 | 620 | 82 | ||
| NM_104424.2 | NP_175945.1 | 2 - 3 | 345 | 82 | ||
| NM_179480.3 | NP_849811.1 | 2 - 3 | 416 | 82 | ||
| GenBank mRNA | AF386945.1 | - | 2 - 3 | 337 | 82 | |
| AK316889.1 | - | 2 - 3 | 322 | 82 | ||
| AY081501.1 | - | 1 - 2 | 215 | 82 | ||
More Information
Set Membership: |
|

CPN60B
Genomic Map