Assay Details
| Gene Symbol: CPN60B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333710.1 | NP_001320639.1 | 6 - 7 | 1259 | 94 | |
| NM_001333711.1 | NP_001320638.1 | 6 - 7 | 1242 | 94 | ||
| NM_001333712.1 | NP_001320637.1 | 6 - 7 | 1225 | 94 | ||
| NM_104424.2 | NP_175945.1 | 6 - 7 | 950 | 94 | ||
| NM_179480.3 | NP_849811.1 | 6 - 7 | 1021 | 94 | ||
| GenBank mRNA | AF386945.1 | - | 6 - 7 | 942 | 94 | |
| AK316889.1 | - | 6 - 7 | 927 | 94 | ||
| AY081501.1 | - | 5 - 6 | 820 | 94 | ||
More Information
Set Membership: |
|

CPN60B
Genomic Map