Assay Details
Gene Symbol: BRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124029.2 | NP_001117501.1 | 2 | 2878 | 79 | |
NM_104437.3 | NP_175957.1 | 3 | 2770 | 79 | ||
GenBank mRNA | FJ708660.1 | - | 1 | 2346 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
