Assay Details
Gene Symbol: AHG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333740.1 | NP_001319244.1 | 1 - 2 | 448 | 83 | |
NM_001333741.1 | NP_001322687.1 | 1 - 2 | 451 | 83 | ||
NM_001333742.1 | NP_001322686.1 | 1 - 2 | 451 | 83 | ||
NM_104464.7 | NP_175983.5 | 1 - 2 | 451 | 83 | ||
GenBank mRNA | AB194966.1 | - | 1 - 2 | 370 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001333740.1, NM_001333741.1, NM_001333742.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
