Assay Details
| Gene Symbol: TIM50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198317.1 | NP_001185246.1 | 2 - 3 | 514 | 88 | |
| NM_104467.5 | NP_175986.2 | 2 - 3 | 501 | 88 | ||
| GenBank mRNA | AY072140.1 | - | 2 - 3 | 498 | 88 | |
| AY122975.1 | - | 2 - 3 | 404 | 88 | ||
More Information
Set Membership: |
|

TIM50
Genomic Map