Assay Details
| Gene Symbol: TIM50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198317.1 | NP_001185246.1 | 8 - 9 | 1090 | 64 | |
| NM_104467.5 | NP_175986.2 | 8 - 9 | 1077 | 64 | ||
| GenBank mRNA | AY072140.1 | - | 8 - 9 | 1074 | 64 | |
| AY122975.1 | - | 8 - 9 | 980 | 64 | ||
More Information
Set Membership: |
|

TIM50
Genomic Map