Assay Details
| Gene Symbol: ZIP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104468.3 | NP_564703.1 | 1 - 2 | 718 | 107 | |
| GenBank mRNA | AF367763.1 | - | 1 - 2 | 495 | 107 | |
| AF410328.1 | - | 1 - 2 | 498 | 107 | ||
| AY093733.1 | - | 1 - 2 | 471 | 107 | ||
More Information
Set Membership: |
|

ZIP11
Genomic Map