Assay Details
| Gene Symbol: HAC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333753.1 | NP_001319247.1 | 9 - 10 | 3162 | 85 | |
| NM_104475.3 | NP_564706.1 | 10 - 11 | 3182 | 85 | ||
More Information
Set Membership: |
|

HAC4
Genomic Map