Assay Details
| Gene Symbol: HAC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333753.1 | NP_001319247.1 | 14 - 15 | 4301 | 71 | |
| NM_104475.3 | NP_564706.1 | 15 - 16 | 4321 | 71 | ||
More Information
Set Membership: |
|

HAC4
Genomic Map