Assay Details
| Gene Symbol: ATP-PRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104590.4 | NP_176105.1 | 4 - 5 | 780 | 98 | |
| GenBank mRNA | AB025249.1 | - | 4 - 5 | 535 | 98 | |
| AY093018.1 | - | 4 - 5 | 554 | 98 | ||
| AY128941.1 | - | 4 - 5 | 530 | 98 | ||
More Information
Set Membership: |
|

ATP-PRT1
Genomic Map