Assay Details
| Gene Symbol: BPEp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104657.4 | NP_564749.1 | 4 - 5 | 849 | 125 | |
| NM_179498.3 | NP_849829.1 | 4 - 5 | 793 | 125 | ||
| GenBank mRNA | AM269753.1 | - | 4 - 5 | 726 | 125 | |
| AM269754.1 | - | 4 - 5 | 726 | 125 | ||
| BT003964.1 | - | 4 - 5 | 750 | 125 | ||
| BT005064.1 | - | 4 - 5 | 590 | 125 | ||
More Information
Set Membership: |
|

BPEp
Genomic Map