Assay Details
| Gene Symbol: TH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104666.4 | NP_176182.1 | 2 - 3 | 348 | 118 | |
| GenBank mRNA | AK228099.1 | - | 2 - 3 | 324 | 118 | |
| BT003140.1 | - | 2 - 3 | 260 | 118 | ||
More Information
Set Membership: |
|

TH7
Genomic Map