Assay Details
| Gene Symbol: AT1G60080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104700.5 | NP_176216.1 | 7 - 8 | 745 | 90 | |
| GenBank mRNA | AK227971.1 | - | 7 - 8 | 683 | 90 | |
| BT004614.1 | - | 6 - 7 | 534 | 90 | ||
More Information
Set Membership: |
|

AT1G60080
Genomic Map