Assay Details
| Gene Symbol: ULP1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104712.5 | NP_176228.3 | 11 - 12 | 1676 | 119 | |
| GenBank mRNA | DQ304543.1 | - | 11 - 12 | 1433 | 119 | |
More Information
Set Membership: |
|

ULP1D
Genomic Map