Assay Details
| Gene Symbol: ULP1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104712.5 | NP_176228.3 | 2 - 3 | 627 | 75 | |
| GenBank mRNA | DQ304543.1 | - | 2 - 3 | 384 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ULP1D| Gene Symbol: ULP1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104712.5 | NP_176228.3 | 2 - 3 | 627 | 75 | |
| GenBank mRNA | DQ304543.1 | - | 2 - 3 | 384 | 75 | |
Set Membership: |
|