Assay Details
| Gene Symbol: AT1G60420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104729.4 | NP_564756.1 | 3 - 4 | 1290 | 97 | |
| GenBank mRNA | AY065433.1 | - | 3 - 4 | 1110 | 97 | |
| AY117231.1 | - | 3 - 4 | 1066 | 97 | ||
More Information
Set Membership: |
|

AT1G60420
Genomic Map