Assay Details
| Gene Symbol: AT1G60590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104742.4 | NP_564758.2 | 6 - 7 | 1425 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G60590| Gene Symbol: AT1G60590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104742.4 | NP_564758.2 | 6 - 7 | 1425 | 116 | |
Set Membership: |
|