Assay Details
| Gene Symbol: ACLA-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333917.1 | NP_001320572.1 | 1 - 2 | 264 | 85 | |
| NM_104764.4 | NP_176280.1 | 2 - 3 | 216 | 85 | ||
More Information
Set Membership: |
|

ACLA-2
Genomic Map