Assay Details
| Gene Symbol: SNRK2.10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104774.6 | NP_176290.1 | 2 - 3 | 520 | 70 | |
| NM_179503.3 | NP_849834.1 | 3 - 4 | 347 | 70 | ||
| GenBank mRNA | AY072340.1 | - | 3 - 4 | 343 | 70 | |
| BT002546.1 | - | 2 - 3 | 195 | 70 | ||
More Information
Set Membership: |
|

SNRK2.10
Genomic Map