Assay Details
| Gene Symbol: SCRL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104779.4 | NP_564768.1 | 1 - 2 | 138 | 69 | |
| GenBank mRNA | AK220784.1 | - | 1 - 2 | 138 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCRL6| Gene Symbol: SCRL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104779.4 | NP_564768.1 | 1 - 2 | 138 | 69 | |
| GenBank mRNA | AK220784.1 | - | 1 - 2 | 138 | 69 | |
Set Membership: |
|