Assay Details
| Gene Symbol: EDA16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124052.2 | NP_001117524.1 | 4 - 5 | 1634 | 103 | |
| NM_001124053.2 | NP_001117525.1 | 4 - 5 | 1963 | 103 | ||
| NM_104795.4 | NP_176309.2 | 4 - 5 | 1935 | 103 | ||
More Information
Set Membership: |
|

EDA16
Genomic Map