Assay Details
Gene Symbol: MLO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198352.1 | NP_001185281.1 | 10 - 11 | 1160 | 90 | |
NM_001198354.1 | NP_001185283.1 | 10 - 11 | 1162 | 90 | ||
NM_001333983.1 | NP_001322619.1 | 10 - 11 | 1380 | 90 | ||
NM_001333987.1 | NP_001322616.1 | 10 - 11 | 1404 | 90 | ||
NM_001333989.1 | NP_001322618.1 | 10 - 11 | 1380 | 90 | ||
NM_104836.2 | NP_176350.1 | 10 - 11 | 1404 | 90 | ||
GenBank mRNA | AF369567.2 | - | 10 - 11 | 1159 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
