Assay Details
Gene Symbol: MLO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198352.1 | NP_001185281.1 | 11 - 12 | 1189 | 81 | |
NM_001198354.1 | NP_001185283.1 | 11 - 12 | 1191 | 81 | ||
NM_001333983.1 | NP_001322619.1 | 11 - 12 | 1409 | 81 | ||
NM_001333987.1 | NP_001322616.1 | 11 - 12 | 1433 | 81 | ||
NM_001333989.1 | NP_001322618.1 | 11 - 12 | 1409 | 81 | ||
NM_104836.2 | NP_176350.1 | 11 - 12 | 1433 | 81 | ||
GenBank mRNA | AF369567.2 | - | 11 - 12 | 1188 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
