Assay Details
Gene Symbol: MLO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198352.1 | NP_001185281.1 | 4 - 5 | 569 | 91 | |
NM_001198354.1 | NP_001185283.1 | 4 - 5 | 571 | 91 | ||
NM_001333983.1 | NP_001322619.1 | 4 - 5 | 789 | 91 | ||
NM_001333984.1 | NP_001319285.1 | 4 - 5 | 734 | 91 | ||
NM_001333985.1 | NP_001322617.1 | 4 - 5 | 813 | 91 | ||
NM_001333986.1 | NP_001322615.1 | 4 - 5 | 813 | 91 | ||
NM_001333987.1 | NP_001322616.1 | 4 - 5 | 813 | 91 | ||
NM_001333988.1 | NP_001322614.1 | 4 - 5 | 813 | 91 | ||
NM_001333989.1 | NP_001322618.1 | 4 - 5 | 789 | 91 | ||
NM_104836.2 | NP_176350.1 | 4 - 5 | 813 | 91 | ||
GenBank mRNA | AF369567.2 | - | 4 - 5 | 568 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
