Assay Details
| Gene Symbol: AT1G61770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104859.3 | NP_176370.2 | 1 - 2 | 233 | 102 | |
| GenBank mRNA | BT002337.1 | - | 1 - 2 | 124 | 102 | |
More Information
Set Membership: |
|

AT1G61770
Genomic Map