Assay Details
| Gene Symbol: CDS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104923.4 | NP_176433.2 | 11 - 12 | 1497 | 64 | |
| GenBank mRNA | X94306.1 | - | 1428 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDS1| Gene Symbol: CDS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104923.4 | NP_176433.2 | 11 - 12 | 1497 | 64 | |
| GenBank mRNA | X94306.1 | - | 1428 | 64 | ||
Set Membership: |
|