Assay Details
| Gene Symbol: AT1G62590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104937.3 | NP_176447.1 | 1 - 2 | 1986 | 116 | |
| Gene Symbol: AT1G63330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334080.1 | NP_001322078.1 | 1 - 2 | 1711 | 116 | |
More Information
Set Membership: |
|

AT1G62590
Genomic Map