Assay Details
| Gene Symbol: SCO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104952.2 | NP_564801.1 | 3 - 4 | 2305 | 69 | |
| GenBank mRNA | AY142646.1 | - | 3 - 4 | 2179 | 69 | |
More Information
Set Membership: |
|

SCO1
Genomic Map