Assay Details
| Gene Symbol: NRPD1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198369.1 | NP_001185298.1 | 14 - 15 | 3335 | 88 | |
| NM_001334061.1 | NP_001320709.1 | 13 - 14 | 3525 | 88 | ||
| NM_001334062.1 | NP_001320708.1 | 14 - 15 | 3335 | 88 | ||
| NM_001334063.1 | NP_001320710.1 | 5 - 6 | 2422 | 88 | ||
| NM_104980.4 | NP_176490.2 | 13 - 14 | 3500 | 88 | ||
| GenBank mRNA | AY826515.1 | - | 12 - 13 | 3226 | 88 | |
| DQ020657.1 | - | 13 - 14 | 3500 | 88 | ||
More Information
Set Membership: |
|

NRPD1A
Genomic Map