Assay Details
| Gene Symbol: NRPD1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198369.1 | NP_001185298.1 | 16 - 17 | 3776 | 71 | |
| NM_001334063.1 | NP_001320710.1 | 7 - 8 | 2863 | 71 | ||
| NM_104980.4 | NP_176490.2 | 15 - 16 | 3941 | 71 | ||
| GenBank mRNA | AY826515.1 | - | 14 - 15 | 3667 | 71 | |
| DQ020657.1 | - | 15 - 16 | 3941 | 71 | ||
More Information
Set Membership: |
|

NRPD1A
Genomic Map