Assay Details
| Gene Symbol: NRPD1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198369.1 | NP_001185298.1 | 19 - 20 | 4276 | 62 | |
| NM_001334061.1 | NP_001320709.1 | 18 - 19 | 4424 | 62 | ||
| NM_001334062.1 | NP_001320708.1 | 18 - 19 | 4076 | 62 | ||
| NM_001334063.1 | NP_001320710.1 | 10 - 11 | 3363 | 62 | ||
| NM_104980.4 | NP_176490.2 | 18 - 19 | 4441 | 62 | ||
| GenBank mRNA | AY826515.1 | - | 17 - 18 | 4167 | 62 | |
| DQ020657.1 | - | 18 - 19 | 4441 | 62 | ||
More Information
Set Membership: |
|

NRPD1A
Genomic Map