Assay Details
| Gene Symbol: NRPD1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198369.1 | NP_001185298.1 | 10 - 11 | 2840 | 70 | |
| NM_001334061.1 | NP_001320709.1 | 9 - 10 | 3030 | 70 | ||
| NM_001334062.1 | NP_001320708.1 | 10 - 11 | 2840 | 70 | ||
| NM_001334063.1 | NP_001320710.1 | 1 - 2 | 1927 | 70 | ||
| NM_104980.4 | NP_176490.2 | 9 - 10 | 3005 | 70 | ||
| GenBank mRNA | AY826515.1 | - | 8 - 9 | 2731 | 70 | |
| DQ020657.1 | - | 9 - 10 | 3005 | 70 | ||
More Information
Set Membership: |
|

NRPD1A
Genomic Map