Assay Details
| Gene Symbol: SEC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117282.3 | NP_567388.1 | 13 - 14 | 1412 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEC1B| Gene Symbol: SEC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117282.3 | NP_567388.1 | 13 - 14 | 1412 | 96 | |
Set Membership: |
|