Assay Details
| Gene Symbol: AIR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336523.1 | NP_001324697.1 | 14 - 15 | 2622 | 105 | |
| NM_129022.8 | NP_181015.7 | 14 - 15 | 2433 | 105 | ||
More Information
Set Membership: |
|

AIR9
Genomic Map