Assay Details
| Gene Symbol: AIR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336523.1 | NP_001324697.1 | 29 - 30 | 4595 | 87 | |
| NM_129022.8 | NP_181015.7 | 29 - 30 | 4406 | 87 | ||
More Information
Set Membership: |
|

AIR9
Genomic Map