Assay Details
| Gene Symbol: AT2G34890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129042.1 | NP_181035.1 | 15 - 16 | 1449 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT2G34890| Gene Symbol: AT2G34890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129042.1 | NP_181035.1 | 15 - 16 | 1449 | 106 | |
Set Membership: |
|