Assay Details
| Gene Symbol: ATL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129052.3 | NP_181045.1 | 1 | 1556 | 131 | |
| GenBank mRNA | AK228813.1 | - | 1 | 1205 | 131 | |
| BT015733.1 | - | 1 | 1186 | 131 | ||
More Information
Set Membership: |
|

ATL9
Genomic Map