Assay Details
| Gene Symbol: UBC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334127.1 | NP_001321725.1 | 4 - 5 | 686 | 65 | |
| NM_105055.3 | NP_564817.2 | 5 - 6 | 544 | 65 | ||
| GenBank mRNA | AK230439.1 | - | 5 - 6 | 481 | 65 | |
| AY074347.1 | - | 5 - 6 | 501 | 65 | ||
| AY114061.1 | - | 5 - 6 | 422 | 65 | ||
| DQ027020.1 | - | 5 - 6 | 422 | 65 | ||
More Information
Set Membership: |
|

UBC5
Genomic Map