Assay Details
Gene Symbol: SPO11-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334143.1 | NP_001320320.1 | 2 - 3 | 197 | 156 | |
NM_105072.3 | NP_176582.2 | 2 - 3 | 197 | 156 | ||
GenBank mRNA | AJ251990.1 | - | 2 - 3 | 186 | 156 | |
DQ446392.1 | - | 2 - 3 | 163 | 156 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
