Assay Details
| Gene Symbol: RHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105119.5 | NP_176625.1 | 1 - 2 | 311 | 69 | |
| GenBank mRNA | BT011226.1 | - | 1 - 2 | 223 | 69 | |
| BT012154.1 | - | 1 - 2 | 184 | 69 | ||
More Information
Set Membership: |
|

RHD1
Genomic Map