Assay Details
| Gene Symbol: RHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105119.5 | NP_176625.1 | 4 - 5 | 596 | 70 | |
| GenBank mRNA | BT011226.1 | - | 4 - 5 | 508 | 70 | |
| BT012154.1 | - | 4 - 5 | 469 | 70 | ||
More Information
Set Membership: |
|

RHD1
Genomic Map